siRNA Details
| Gene Symbol: Loxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164311.1 | NP_001157783.1 | 14 | 2239 | ||
| NM_053083.3 | NP_444313.3 | 14 | 2236 | |||
| GenBank mRNA | AF338440.1 | - | 13 | 2131 | ||
| AK014331.1 | - | 14 | 2238 | |||
| BC068307.1 | - | 14 | 2178 | 30059745 | ||

Loxl4
Genomic Map