siRNA Details
| Gene Symbol: Lrrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146021.1 | NP_001139493.1 | 3 | 530 | ||
| NM_026253.4 | NP_080529.1 | 3 | 530 | |||
| GenBank mRNA | AK005666.1 | - | 3 | 470 | ||
| AK006063.1 | - | 3 | 467 | |||
| AK014952.1 | - | 3 | 495 | |||
| AK015365.1 | - | 3 | 534 | |||
| AK029909.1 | - | 3 | 495 | |||
| AY775400.1 | - | 3 | 599 | |||
| BC048444.1 | - | 3 | 515 | |||
| BC061072.1 | - | 1 | 258 | 6771444 | ||

Lrrc18
Genomic Map