siRNA Details
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026254.2 | NP_080530.2 | 8 | 946 | ||
GenBank mRNA | AK155142.1 | - | 8 | 967 | ||
BC031706.1 | - | 8 | 946 | 4194266 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026254.2 | NP_080530.2 | 8 | 946 | ||
GenBank mRNA | AK155142.1 | - | 8 | 967 | ||
BC031706.1 | - | 8 | 946 | 4194266 |