siRNA Details
| Gene Symbol: 4930519G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026263.2 | NP_080539.1 | 5 | 489 | ||
| GenBank mRNA | AK015843.1 | - | 5 | 491 | ||
| AK015857.1 | - | 5 | 540 | |||
| AK161377.1 | - | 3 | 1390 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930519G04Rik| Gene Symbol: 4930519G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026263.2 | NP_080539.1 | 5 | 489 | ||
| GenBank mRNA | AK015843.1 | - | 5 | 491 | ||
| AK015857.1 | - | 5 | 540 | |||
| AK161377.1 | - | 3 | 1390 | |||