siRNA Details
| Gene Symbol: Ube2r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026275.4 | NP_080551.1 | 4 | 534 | ||
| GenBank mRNA | AJ240086.1 | - | 4 | 493 | ||
| AK003550.1 | - | 4 | 554 | |||
| AK007517.1 | - | 4 | 668 | |||
| AK075703.1 | - | 4 | 544 | |||
| AK075714.1 | - | 4 | 535 | |||
| AK182726.1 | - | 1 | 129 | |||
| AK183933.1 | - | 1 | 129 | |||
| AK217134.1 | - | 1 | 129 | |||
| BC011112.1 | - | 4 | 542 | 3982808 | ||

Ube2r2
Genomic Map