siRNA Details
| Gene Symbol: Bend5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026279.3 | NP_080555.1 | 5, 6 | 1274 | ||
| GenBank mRNA | AK075885.1 | - | 5, 6 | 1274 | ||
| AK156708.1 | - | 5, 6 | 1176 | |||
| AK167425.1 | - | 5, 6 | 1094 | |||
| BC037633.1 | - | 5, 6 | 1107 | |||
| BC051512.1 | - | 3, 4 | 695 | 1383740 | ||

Bend5
Genomic Map