siRNA Details
| Gene Symbol: Mxra7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026280.3 | NP_080556.1 | 4 | 511 | ||
| GenBank mRNA | AK012601.1 | - | 4 | 483 | ||
| AK075801.1 | - | 4 | 512 | |||
| BC147320.1 | - | 4 | 592 | 9056373 | ||
| BC147321.1 | - | 4 | 592 | 9056376 | ||

Mxra7
Genomic Map