siRNA Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026281.2 | NP_080557.2 | 5 | 702 | ||
| GenBank mRNA | AK010720.1 | - | 5 | 545 | ||
| AK154633.1 | - | 5 | 702 | |||
| AK171111.1 | - | 5 | 679 | |||
| BC023389.1 | - | 5 | 641 | |||
| BC049960.1 | - | 3 | 338 | |||
| DQ104329.1 | - | 5 | 650 | |||

Tm7sf3
Genomic Map