siRNA Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026281.2 | NP_080557.2 | 12 | 1606 | ||
| GenBank mRNA | AK010720.1 | - | 12 | 1449 | ||
| AK154633.1 | - | 12 | 1606 | |||
| AK171111.1 | - | 12 | 1583 | |||
| AK214598.1 | - | 2 | 108 | |||
| AK217374.1 | - | 2 | 108 | |||
| BC023389.1 | - | 12 | 1545 | |||
| BC049960.1 | - | 10 | 1242 | |||
| BC057900.1 | - | 7 | 708 | |||
| DQ104329.1 | - | 12 | 1554 | |||

Tm7sf3
Genomic Map