siRNA Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026281.2 | NP_080557.2 | 6 | 822 | ||
| GenBank mRNA | AK010720.1 | - | 6 | 665 | ||
| AK154633.1 | - | 6 | 822 | |||
| AK171111.1 | - | 6 | 799 | |||
| AK181433.1 | - | 2 | 122 | |||
| BC023389.1 | - | 6 | 761 | |||
| BC049960.1 | - | 4 | 458 | |||
| DQ104329.1 | - | 6 | 770 | |||

Tm7sf3
Genomic Map