siRNA Details
| Gene Symbol: Spc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026282.5 | NP_080558.1 | 4 | 445 | ||
| GenBank mRNA | AK011728.1 | - | 4 | 463 | ||
| AK031680.1 | - | 3 | 538 | |||
| AK075950.1 | - | 4 | 446 | |||
| AK153185.1 | - | 4 | 463 | |||
| BC086683.1 | - | 4 | 448 | 30676712 | ||
| BC098472.1 | - | 453 | ||||
| BC116238.1 | - | 4 | 446 | 40091783 | ||
| BC116239.1 | - | 4 | 445 | 40091784 | ||

Spc24
Genomic Map