siRNA Details
| Gene Symbol: 4930522H14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199090.1 | NP_001186019.1 | 3 | 273 | ||
| NM_026291.3 | NP_080567.1 | 3 | 250 | |||
| GenBank mRNA | AK005726.1 | - | 3 | 274 | ||
| AK015869.1 | - | 3 | 192 | |||
| AK019777.1 | - | 3 | 251 | |||
| BC125547.1 | - | 3 | 210 | 40130062 | ||
| BC125549.1 | - | 3 | 211 | 40130064 | ||

4930522H14Rik
Genomic Map