siRNA Details
| Gene Symbol: Spaca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026293.3 | NP_080569.1 | 7 | 948 | ||
| GenBank mRNA | AK006187.1 | - | 7 | 949 | ||
| AK016011.1 | - | 7 | 941 | |||
| BC147079.1 | - | 7 | 945 | 9056040 | ||
| BC147080.1 | - | 7 | 945 | 9056039 | ||

Spaca1
Genomic Map