siRNA Details
| Gene Symbol: Spaca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290443.1 | NP_001277372.1 | 6 | 796 | ||
| NM_026293.3 | NP_080569.1 | 6 | 798 | |||
| GenBank mRNA | AK006187.1 | - | 6 | 799 | ||
| AK007270.1 | - | 6 | 770 | |||
| AK016011.1 | - | 6 | 791 | |||
| BC048388.1 | - | 3 | 455 | |||
| BC147079.1 | - | 6 | 795 | 9056040 | ||
| BC147080.1 | - | 6 | 795 | 9056039 | ||

Spaca1
Genomic Map