siRNA Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026295.2 | NP_080571.2 | 6, 7 | 982 | ||
| GenBank mRNA | AK156617.1 | - | 6, 7 | 1011 | ||
| AK186350.1 | - | 2, 3 | 137 | |||
| AK206583.1 | - | 2, 3 | 133 | |||
| BC053435.1 | - | 6, 7 | 982 | 6394578 | ||

Ctdp1
Genomic Map