siRNA Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026295.2 | NP_080571.2 | 7 | 1025 | ||
| GenBank mRNA | AK156617.1 | - | 7 | 1054 | ||
| AK186350.1 | - | 3 | 182 | |||
| AK206583.1 | - | 3 | 176 | |||
| BC053435.1 | - | 7 | 1025 | 6394578 | ||

Ctdp1
Genomic Map