siRNA Details
| Gene Symbol: 4930548H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026296.3 | NP_080572.1 | 1 | 259 | ||
| GenBank mRNA | AK016070.1 | - | 1 | 253 | ||
| AK016690.1 | - | 1 | 260 | |||
| BC057946.1 | - | 1 | 378 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930548H24Rik| Gene Symbol: 4930548H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026296.3 | NP_080572.1 | 1 | 259 | ||
| GenBank mRNA | AK016070.1 | - | 1 | 253 | ||
| AK016690.1 | - | 1 | 260 | |||
| BC057946.1 | - | 1 | 378 | |||