siRNA Details
| Gene Symbol: 4930549C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026300.2 | NP_080576.1 | 2 | 602 | ||
| GenBank mRNA | AK016074.1 | - | 2 | 603 | ||
| BC048442.1 | - | 2 | 592 | 6704947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930549C01Rik| Gene Symbol: 4930549C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026300.2 | NP_080576.1 | 2 | 602 | ||
| GenBank mRNA | AK016074.1 | - | 2 | 603 | ||
| BC048442.1 | - | 2 | 592 | 6704947 | ||