siRNA Details
Gene Symbol: Rnf125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026301.2 | NP_080577.1 | 4, 5 | 479 | ||
GenBank mRNA | AB259692.1 | - | 4, 5 | 510 | ||
AK006569.1 | - | 3, 4 | 307 | |||
AK016103.1 | - | 4, 5 | 676 | |||
AK050395.1 | - | 4, 5 | 480 | |||
BC094290.1 | - | 3, 4 | 276 | |||
BC116883.1 | - | 3, 4 | 348 | 40126202 | ||
BC116887.1 | - | 3, 4 | 348 | 40126206 |
