siRNA Details
| Gene Symbol: Dctn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026302.3 | NP_080578.1 | 12, 13 | 1176 | ||
| GenBank mRNA | AK016059.1 | - | 12, 13 | 1230 | ||
| AK034292.1 | - | 13, 14 | 1234 | |||
| AK141855.1 | - | 13, 14 | 1200 | |||
| AK160813.1 | - | 12, 13 | 1182 | |||
| AK163240.1 | - | 12, 13 | 1198 | |||
| AK168484.1 | - | 12, 13 | 1182 | |||
| BC006677.1 | - | 12, 13 | 1139 | |||
| BC034725.1 | - | 12, 13 | 1176 | 3667031 | ||

Dctn4
Genomic Map