siRNA Details
| Gene Symbol: Luc7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026313.1 | NP_080589.1 | 11 | 1440 | ||
| GenBank mRNA | AK151228.1 | - | 11 | 1488 | ||
| AK151839.1 | - | 11 | 1488 | |||
| BC009092.1 | - | 11 | 1440 | 3157532 | ||
| BC055921.1 | - | 1 | 47 | |||

Luc7l3
Genomic Map