siRNA Details
| Gene Symbol: Aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026316.2 | NP_080592.2 | 4 | 380 | ||
| GenBank mRNA | AF362571.1 | - | 3 | 302 | ||
| AK154500.1 | - | 4 | 426 | |||
| AK154723.1 | - | 4 | 473 | |||
| AK159447.1 | - | 4 | 394 | |||
| AK160106.1 | - | 4 | 397 | |||
| BC046597.1 | - | 4 | 380 | 6312211 | ||

Aldh3b1
Genomic Map