siRNA Details
| Gene Symbol: Rnf149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033135.3 | NP_001028307.2 | 2 | 566 | ||
| GenBank mRNA | AK155360.1 | - | 2 | 625 | ||
| AY155439.1 | - | 2 | 158 | |||
| BC115968.1 | - | 3 | 146 | 40107454 | ||
| BF018252.1 | - | 2 | 315 | |||
| BY743657.1 | - | 2 | 566 | |||

Rnf149
Genomic Map