siRNA Details
| Gene Symbol: Kirrel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190911.1 | NP_001177840.1 | 2, 3 | 500 | ||
| NM_001190913.1 | NP_001177842.1 | 2, 3 | 500 | |||
| NM_001190914.1 | NP_001177843.1 | 3, 4 | 601 | |||
| NM_026324.3 | NP_080600.1 | 2, 3 | 500 | |||
| GenBank mRNA | AK034142.1 | - | 2, 3 | 437 | ||
| AK038385.1 | - | 2, 3 | 251 | |||
| AK045373.1 | - | 2, 3 | 405 | |||
| AK079332.1 | - | 3, 4 | 538 | |||
| AK122566.1 | - | 1, 2 | 14 | |||
| AY169782.1 | - | 2, 3 | 500 | |||

Kirrel3
Genomic Map