siRNA Details
| Gene Symbol: Pcnxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026327.3 | NP_080603.2 | 9 | 2970 | ||
| GenBank mRNA | AK029996.1 | - | 9 | 2927 | ||
| AK136802.1 | - | 9 | 2945 | |||
| AK160852.1 | - | 8 | 2190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcnxl4| Gene Symbol: Pcnxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026327.3 | NP_080603.2 | 9 | 2970 | ||
| GenBank mRNA | AK029996.1 | - | 9 | 2927 | ||
| AK136802.1 | - | 9 | 2945 | |||
| AK160852.1 | - | 8 | 2190 | |||