siRNA Details
| Gene Symbol: Dnajc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001026211.2 | NP_001021382.1 | 4 | 362 | ||
| NM_001286972.1 | NP_001273901.1 | 4 | 362 | |||
| NM_001286973.1 | NP_001273902.1 | 3 | 310 | |||
| NM_026332.4 | NP_080608.3 | 4 | 362 | |||
| GenBank mRNA | AK007866.1 | - | 4 | 330 | ||
| AK008267.1 | - | 4 | 306 | |||
| AK010204.1 | - | 4 | 312 | |||
| AK012051.1 | - | 4 | 314 | |||
| AK076602.1 | - | 321 | ||||
| BC020327.1 | - | 4 | 295 | |||
| BC024335.1 | - | 4 | 270 | 4974988 | ||
| CA469052.1 | - | 4 | 399 | |||
| CJ123658.1 | - | 3 | 267 | |||
| EL962106.1 | - | 4 | 308 | |||

Dnajc19
Genomic Map