siRNA Details
| Gene Symbol: Ribc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026357.2 | NP_080633.2 | 3 | 577 | ||
| GenBank mRNA | AK006801.1 | - | 3 | 574 | ||
| AK016311.1 | - | 3 | 578 | |||
| AK017028.1 | - | 3 | 597 | |||
| BC112373.1 | - | 3 | 502 | 40056860 | ||
| BY716712.1 | - | 3 | 597 | |||

Ribc2
Genomic Map