siRNA Details
| Gene Symbol: 4930578I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026359.3 | NP_080635.2 | 3 | 336 | ||
| GenBank mRNA | AK005778.1 | - | 3 | 333 | ||
| AK016302.1 | - | 345 | ||||
| BC048583.1 | - | 3 | 336 | 6706576 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930578I06Rik| Gene Symbol: 4930578I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026359.3 | NP_080635.2 | 3 | 336 | ||
| GenBank mRNA | AK005778.1 | - | 3 | 333 | ||
| AK016302.1 | - | 345 | ||||
| BC048583.1 | - | 3 | 336 | 6706576 | ||