siRNA Details
| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026360.3 | NP_080636.2 | 9, 10 | 1073 | ||
| GenBank mRNA | AK010310.1 | - | 9, 10 | 1066 | ||
| AK076982.1 | - | 9, 10 | 1058 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx47| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026360.3 | NP_080636.2 | 9, 10 | 1073 | ||
| GenBank mRNA | AK010310.1 | - | 9, 10 | 1066 | ||
| AK076982.1 | - | 9, 10 | 1058 | |||