siRNA Details
| Gene Symbol: Gpatch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026367.4 | NP_080643.4 | 2 | 334 | ||
| GenBank mRNA | AK011727.1 | - | 2 | 319 | ||
| AK017975.1 | - | 2 | 303 | |||
| AK029990.1 | - | 2 | 319 | |||
| AK030026.1 | - | 2 | 333 | |||
| AK032734.1 | - | 2 | 309 | |||
| AK053781.1 | - | 2 | 326 | |||
| AK167826.1 | - | 2 | 338 | |||
| AK169660.1 | - | 2 | 335 | |||
| BC054810.1 | - | 2 | 306 | 6839419 | ||

Gpatch2
Genomic Map