siRNA Details
| Gene Symbol: Caap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC020067.1 | - | 2 | 472 | 3991510 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Caap1| Gene Symbol: Caap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC020067.1 | - | 2 | 472 | 3991510 | |