siRNA Details
| Gene Symbol: Loh12cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170479.1 | NP_001163950.1 | 3, 4 | 476 | ||
| NM_026371.3 | NP_080647.2 | 3, 4 | 618 | |||
| GenBank mRNA | AK007423.1 | - | 3, 4 | 475 | ||
| AK018019.1 | - | 3, 4 | 534 | |||
| AK089648.1 | - | 3, 4 | 482 | |||
| AK151342.1 | - | 3, 4 | 484 | |||
| AK169615.1 | - | 3, 4 | 490 | |||
| BC023059.1 | - | 3, 4 | 577 | 5355560 | ||
| DV652098.1 | - | 3, 4 | 368 | |||
| DV658678.1 | - | 3, 4 | 478 | |||

Loh12cr1
Genomic Map