siRNA Details
| Gene Symbol: Loh12cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170479.1 | NP_001163950.1 | 4 | 845 | ||
| NM_026371.3 | NP_080647.2 | 4 | 987 | |||
| GenBank mRNA | AK007423.1 | - | 4 | 844 | ||
| AK018019.1 | - | 4 | 903 | |||
| AK089648.1 | - | 4 | 851 | |||
| AK151342.1 | - | 4 | 853 | |||
| AK169615.1 | - | 4 | 859 | |||
| BC023059.1 | - | 4 | 946 | 5355560 | ||

Loh12cr1
Genomic Map