siRNA Details
| Gene Symbol: Plxnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026376.3 | NP_080652.2 | 34, 35 | 5786 | ||
| GenBank mRNA | AK129175.1 | - | 34, 35 | 5051 | ||
| AK147513.1 | - | 34, 35 | 5787 | |||
| AY688678.1 | - | 34, 35 | 5746 | |||

Plxnd1
Genomic Map