siRNA Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026378.3 | NP_080654.3 | 9 | 1211 | ||
| GenBank mRNA | AK002439.1 | - | 9 | 1207 | ||
| AK003374.1 | - | 3 | 158 | |||
| AK149963.1 | - | 9 | 1212 | |||
| AK169405.1 | - | 9 | 1209 | |||
| AK171818.1 | - | 9 | 1212 | |||
| BC013483.1 | - | 9 | 1192 | 4242135 | ||
| BC058774.1 | - | 7 | 610 | |||

Dalrd3
Genomic Map