siRNA Details
| Gene Symbol: Dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026378.3 | NP_080654.3 | 10, 11 | 1445 | ||
| GenBank mRNA | AK002439.1 | - | 10, 11 | 1441 | ||
| AK003374.1 | - | 4, 5 | 392 | |||
| AK149963.1 | - | 10, 11 | 1446 | |||
| AK169405.1 | - | 10, 11 | 1443 | |||
| AK171818.1 | - | 10, 11 | 1446 | |||
| AK185660.1 | - | 1, 2 | 38 | |||
| BC013483.1 | - | 10, 11 | 1426 | 4242135 | ||
| BC058774.1 | - | 8, 9 | 844 | |||

Dalrd3
Genomic Map