siRNA Details
| Gene Symbol: Rab39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175122.6 | NP_780331.1 | 2 | 817 | ||
| GenBank mRNA | AB232636.1 | - | 2 | 593 | ||
| AK018257.1 | - | 2 | 801 | |||
| AK020665.1 | - | 2 | 800 | |||
| AK039013.1 | - | 2 | 791 | |||
| AK048981.1 | - | 2 | 791 | |||
| AK160737.1 | - | 2 | 791 | |||
| BC050853.1 | - | 2 | 811 | 6517891 | ||
| BC052472.1 | - | 2 | 742 | 6833461 | ||

Rab39b
Genomic Map