siRNA Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026380.3 | NP_080656.2 | 4, 5 | 625 | ||
| GenBank mRNA | AK044337.1 | - | 4, 5 | 499 | ||
| AK142301.1 | - | 4, 5 | 494 | |||
| BC065056.1 | - | 4, 5 | 558 | 5705033 | ||
| BC094577.1 | - | 3, 4 | 241 | 6852696 | ||

Rgs8
Genomic Map