siRNA Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026380.3 | NP_080656.2 | 6 | 804 | ||
| GenBank mRNA | AK044337.1 | - | 6 | 678 | ||
| AK142301.1 | - | 6 | 673 | |||
| BC065056.1 | - | 6 | 737 | 5705033 | ||
| BC094577.1 | - | 5 | 420 | 6852696 | ||

Rgs8
Genomic Map