siRNA Details
| Gene Symbol: Snrnp48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026382.2 | NP_080658.2 | 4 | 422 | ||
| GenBank mRNA | AK007431.1 | - | 4 | 411 | ||
| AK012916.1 | - | 4 | 417 | |||
| AK018315.1 | - | 4 | 417 | |||
| AK036713.1 | - | 4 | 422 | |||
| AK087789.1 | - | 4 | 417 | |||
| AK172155.1 | - | 4 | 411 | |||
| BC021443.1 | - | 4 | 405 | 5038605 | ||
| BC037073.1 | - | 4 | 412 | |||

Snrnp48
Genomic Map