siRNA Details
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172397.3 | NP_765985.1 | 3, 4 | 229 | ||
GenBank mRNA | AK012581.1 | - | 3, 4 | 203 | ||
AK032430.1 | - | 3, 4 | 230 | |||
AK040486.1 | - | 3, 4 | 230 | |||
AK049809.1 | - | 3, 4 | 199 | |||
AK167349.1 | - | 3, 4 | 199 | |||
AK169829.1 | - | 3, 4 | 200 | |||
AK171794.1 | - | 3, 4 | 508 | |||
BC068130.1 | - | 3, 4 | 173 | 6413206 |
