siRNA Details
| Gene Symbol: Sec14l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144520.2 | NP_653103.1 | 9 | 787 | ||
| GenBank mRNA | BC005759.1 | - | 9 | 787 | 2655022 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec14l2| Gene Symbol: Sec14l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144520.2 | NP_653103.1 | 9 | 787 | ||
| GenBank mRNA | BC005759.1 | - | 9 | 787 | 2655022 | |