siRNA Details
| Gene Symbol: MC5R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005913.2 | NP_005904.1 | 1 | 514 | ||
| GenBank mRNA | BC069153.1 | - | 1 | 293 | 7217013 | |
| BC069545.1 | - | 1 | 292 | 7262232 | ||
| BC095531.1 | - | 1 | 517 | 6971958 | ||
More Information
Set Membership: |
|

MC5R
Genomic Map