siRNA Details
| Gene Symbol: Snap47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144521.2 | NP_653104.1 | 2 | 289 | ||
| GenBank mRNA | AK028053.1 | - | 2 | 305 | ||
| AK049330.1 | - | 2 | 333 | |||
| AK075640.1 | - | 2 | 328 | |||
| AK081883.1 | - | 2 | 305 | |||
| AK144165.1 | - | 2 | 342 | |||
| BC023182.1 | - | 2 | 289 | 4949809 | ||
| BC029025.1 | - | 2 | 299 | 5036036 | ||

Snap47
Genomic Map