siRNA Details
| Gene Symbol: Lce1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026394.3 | NP_080670.2 | 2 | 675 | ||
| GenBank mRNA | AK003518.1 | - | 2 | 676 | ||
| BC060283.1 | - | 2 | 659 | |||
| BC100541.1 | - | 1 | 643 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lce1f| Gene Symbol: Lce1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026394.3 | NP_080670.2 | 2 | 675 | ||
| GenBank mRNA | AK003518.1 | - | 2 | 676 | ||
| BC060283.1 | - | 2 | 659 | |||
| BC100541.1 | - | 1 | 643 | |||