siRNA Details
| Gene Symbol: Mrpl57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026401.2 | NP_080677.1 | 2 | 177 | ||
| GenBank mRNA | AB049958.1 | - | 2 | 138 | ||
| AK003130.1 | - | 2 | 173 | |||
| AK010282.1 | - | 2 | 182 | |||
| AK011558.1 | - | 2 | 178 | |||
| BC049555.1 | - | 172 | 6772707 | |||
| BC055723.1 | - | 2 | 118 | 6411308 | ||

Mrpl57
Genomic Map