siRNA Details
| Gene Symbol: MCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085377.1 | NP_001078846.1 | 18 | 3263 | ||
| NM_002387.2 | NP_002378.1 | 16 | 2807 | |||
| XM_005271991.2 | XP_005272048.1 | 17 | 2907 | |||
| XM_017009473.1 | XP_016864962.1 | 19 | 2928 | |||
| XM_017009474.1 | XP_016864963.1 | 17 | 5593 | |||
| GenBank mRNA | AK128596.1 | - | 19 | 3275 | ||
| BC009279.2 | - | 17 | 2807 | 4111706 | ||
| BC018919.2 | - | 17 | 2807 | 4131685 | ||
| BX537952.1 | - | 17 | 2278 | |||
| BX647710.1 | - | 19 | 2737 | |||
| M62397.1 | - | 17 | 2497 | |||

MCC
Genomic Map