siRNA Details
| Gene Symbol: Cdca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026410.3 | NP_080686.1 | 5 | 763 | ||
| GenBank mRNA | AK010540.1 | - | 5 | 730 | ||
| AK011701.1 | - | 5 | 763 | |||
| AK016305.1 | - | 5 | 877 | |||
| AK199612.1 | - | 1 | 70 | |||
| BC029626.1 | - | 4 | 657 | |||
| BC052904.1 | - | 5 | 761 | 30065848 | ||

Cdca5
Genomic Map