siRNA Details
| Gene Symbol: Ppp6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024209.2 | NP_077171.1 | 6 | 599 | ||
| GenBank mRNA | AK002764.1 | - | 6 | 561 | ||
| AK009104.1 | - | 6 | 579 | |||
| BC002223.1 | - | 6 | 624 | 3491111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp6c| Gene Symbol: Ppp6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024209.2 | NP_077171.1 | 6 | 599 | ||
| GenBank mRNA | AK002764.1 | - | 6 | 561 | ||
| AK009104.1 | - | 6 | 579 | |||
| BC002223.1 | - | 6 | 624 | 3491111 | ||