siRNA Details
| Gene Symbol: Rgs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026418.2 | NP_080694.1 | 4 | 434 | ||
| GenBank mRNA | AK009283.1 | - | 4 | 435 | ||
| AK009686.1 | - | 4 | 436 | |||
| AK017314.1 | - | 4 | 435 | |||
| AK088009.1 | - | 4 | 433 | |||
| BC109178.1 | - | 4 | 419 | 40049527 | ||
| BC109179.1 | - | 4 | 424 | |||

Rgs10
Genomic Map